Learn More
GAP-43 Antibody (CL10855) [DyLight 350], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-21182UV
Product Code. 30308063
  • 737.00 € / 0.10mL
Description
GAP-43 Monoclonal antibody specifically detects GAP-43 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GAP-43
Monoclonal
DyLight 350
Axonal membrane protein GAP-43, B-50, calmodulin-binding protein P-57, growth associated protein 43, Growth-associated protein 43, nerve growth-related peptide GAP43, Neural phosphoprotein B-50, neuromodulin, neuron growth-associated protein 43, PP46, protein F1
This antibody was developed using a synthetic peptide derived from P17677, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer, Cellular Markers, Growth and Development, Neuronal Cell Markers, Neuroscience, Phospho Specific
2596
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL10855
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified