missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAP-43 Antibody (S01-4F6), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20078-100ul
This item is not returnable.
View return policy
Description
GAP-43 Monoclonal antibody specifically detects GAP-43 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| GAP-43 | |
| Monoclonal | |
| 0.3 mg/mL | |
| Western Blot, Immunohistochemistry 1:50, Immunohistochemistry-Paraffin | |
| Axonal membrane protein GAP-43, B-50, calmodulin-binding protein P-57, growth associated protein 43, Growth-associated protein 43, nerve growth-related peptide GAP43, Neural phosphoprotein B-50, neuromodulin, neuron growth-associated protein 43, PP46, protein F1 | |
| A synthetic peptide of human GAP-43 (Uniprot # P17677) | |
| 100 μg | |
| Cancer, Cellular Markers, Growth and Development, Neuronal Cell Markers, Neuroscience, Phospho Specific | |
| 2596 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| S01-4F6 | |
| Unconjugated | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction