missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GAS1 Monoclonal Antibody (309325)
GREENER_CHOICE

Product Code. 30536048
Change view
Click to view available options
Quantity:
500 μg
Unit Size:
500µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30536048 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30536048 Supplier Invitrogen™ Supplier No. MA561192

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

Reconstitute at 0.5 mg/mL in sterile PBS.

Growth arrest-specific 1 plays a role in growth suppression. GAS1 blocks entry to S phase and prevents cycling of normal and transformed cells. Gas1 is a putative tumor suppressor gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GAS1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 309325
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with trehalose and no preservative
Gene GAS1
Gene Accession No. P54826
Gene Alias AW554192; GAS1; Gas-1; growth arrest specific 1; growth arrest-specific 1; Growth arrest-specific gene-1; growth arrest-specific protein 1
Gene Symbols GAS1
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Gas1, Leu40-Ser318.
Purification Method Protein A/G
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2619
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.