missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAS2L2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
280.00 € - 624.00 €
Specifications
| Antigen | GAS2L2 |
|---|---|
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:5000 - 1:10000 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18462292
|
Novus Biologicals
NBP2-32029-25ul |
25 μL |
280.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18136182
|
Novus Biologicals
NBP2-32029 |
0.1 mL |
624.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GAS2L2 Polyclonal specifically detects GAS2L2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| GAS2L2 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| Q8NHY3 | |
| 246176 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: TPGRSFPGATSGSPRTELGRDPIPLRAVTVDLAGSTHGDCSVEVRQEDQQLDIQVMAEARESWDLGLQEQEGRYTPLPLGGNKEQAIYCSLEEEILGNMKLLEVRSACPQGTRSGVIPRSGVYIPRLAGQWPEPGGPYDKAIQELAQGS | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:5000 - 1:10000 | |
| Polyclonal | |
| Rabbit | |
| Cell Cycle and Replication | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| GAR17Growth arrest-specific protein 2-like 2, GAS2-like protein 2, GAS2-related protein on chromosome 17, growth arrest-specific 2 like 2 | |
| GAS2L2 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title