missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GAS6 Monoclonal Antibody (100118)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561974
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
This gene product is a gamma-carboxyglutamic acid (Gla)-containing protein thought to be involved in the stimulation of cell proliferation, and may play a role in thrombosis. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
| GAS6 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q14393 | |
| GAS6 | |
| Mouse myeloma cell line NS0-derived recombinant human Gas6, Asp118-Ala678. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| ELISA | |
| 100118 | |
| Unconjugated | |
| GAS6 | |
| AXL receptor tyrosine kinase ligand; AXL stimulatory factor; AXLLG; AXSF; GAS 6; GAS6; GAS-6; GPF; growth arrest specific 6; growth arrest-specific 6; growth arrest-specific protein 6; growth-potentiating factor | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 2621 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction