missing translation for 'onlineSavingsMsg'
Learn More

GATA-6 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

369.00 € - 593.00 €

Specifications

Antigen GATA-6
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18209814
View Documents
Novus Biologicals
NBP2-55937
100 μL
593.00 €
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18684329
View Documents
Novus Biologicals
NBP2-55937-25ul
25 μL
369.00 €
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

GATA-6 Polyclonal specifically detects GATA-6 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

GATA-6
Polyclonal
Rabbit
Stem Cell Markers
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
2627
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:INKSKTCSGNSNNSIPMTPTSTSSNSDDCSKNTSPTTQPTASGAGAPVMTGAGE
Primary
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Immunocytochemistry, Immunofluorescence
Unconjugated
RUO
Human
GATA binding protein 6, GATA-binding factor 6, GATA-binding protein 6, transcription factor GATA-6
GATA6
IgG
Affinity Purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.