missing translation for 'onlineSavingsMsg'
Learn More

Gatifloxacin, MedChemExpress

Product Code. 16471059
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16471059 1 g 1g
16456757 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16471059 Supplier Medchem Express Supplier No. HY10581
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Gatifloxacin (AM-1155; BMS-206584; PD135432) is a potent fluoroquinolone antibiotic with broad-spectrum antibacterial activity. Gatifloxacin inhibits bacterial type II topoisomerases (IC50=13.8 μg/ml for S. aureus topoisomerase IV) and E. coli DNA gyrase (IC50=0.109 μg/ml). Gatifloxacin can be used to treat bacterial conjunctivitis in vivo.

Gatifloxacin (AM-1155

Specifications

Chemical Name or Material Gatifloxacin
Physical Form Solid
CAS 112811-59-3
Color White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C19H22FN3O4
For Use With (Application) COVID-19-anti-virus
Synonym AM-1155 BMS-206584 PD135432
Solubility Information DMSO : 2 mg/mL (5.33 mM; Need ultrasonic) ∣H2O : 1 mg/mL (2.66 mM; Need ultrasonic)
SMILES CC1NCCN(C2=C(F)C=C3C(N(C4CC4)C=C(C(O)=O)C3=O)=C2OC)C1
Molecular Weight (g/mol) 375.39
Formula Weight 375.39
Percent Purity 99.37%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.