missing translation for 'onlineSavingsMsg'
Learn More

GB 23200.113-2018 Group B 105 Pesticides 10 ug/ml in Ethyl acetate, Dr. Ehrenstorfer

Product Code. 30323254
Change view
Click to view available options
Quantity:
1.5 mL
Unit Size:
1.5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30323254 1.5 mL 1.5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30323254 Supplier Dr. Ehrenstorfer Supplier No. DREA50000093EA
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (±)-Fenpropathrin∣(EZ)-Tetrachlorvinphos∣2,3,5,6-Tetrachloronitrobenzene∣Acrinathrin∣Aldrin∣Ametryn∣Atraton∣Atrazine∣Azinphos-ethyl∣Beflubutamid∣Benalaxyl∣Benfluralin∣Bifenox∣Biphenyl∣Bromophos-ethyl∣Butachlor∣Butamifos∣Carbofuran∣Chlorfenson∣Chlorfenvinphos∣Chlorobenzilate∣Chloroneb∣Chlorpropham∣Chlorpyrifos∣Chlorpyrifos-methyl∣Clomazone∣Coumaphos∣Cyfluthrin∣Cyproconazole∣Cyprodinil∣Desmetryn∣Diazinon∣Diclofop methyl∣Dicrotophos∣Dieldrin∣Difenoconazole∣Diniconazole∣Diphenylamine∣Dipropetryn∣Ethiolate∣Ethion∣Ethofumesate∣Etoxazole∣Etridiazole∣Etrimfos∣Famphur∣Fenbuconazole∣Fenchlorphos∣Fenitrothion∣Fenobucarb∣Fipronil∣Fluazifop-butyl∣Flucythrinate∣Fludioxonil∣Fluorodifen∣Fluquinconazole∣Hexaconazole∣Iprodione∣Isazofos∣Isocarbofos∣Isofenphos-oxon∣Isoprothiolane∣lambda-Cyhalothrin∣Leptophos∣Malaoxon∣Malathion∣Mefenacet∣Methidathion∣Methoprene∣Methoxychlor∣Monolinuron∣Myclobutanil∣Naled∣Napropamide∣Nitrofen∣Omethoate∣Oxadixyl∣Paclobutrazol∣Parathion-methyl∣Pendimethalin∣Pentachloroaniline∣Phosalone∣Phosfolan∣Phosmet∣Phosphamidon∣Pirimiphos-ethyl∣Procymidone∣Profenofos∣Prometryn∣Propanil∣Propiconazole∣Propyzamide∣Prothiophos∣Pyridaphenthion∣Pyrimethanil∣Quintozene∣Simazine∣tau-Fluvalinate∣Terbuthylazine∣Terbutryn∣Tetraconazole∣Thionazin∣Tolclofos-methyl∣Trans-Chlordane (Gamma Isomer)∣Trichloronate
CAS 39515-41-8, 961-11-5, 117-18-0, 101007-06-1, 309-00-2, 834-12-8
Analyte or Component Names (±)-Fenpropathrin∣(EZ)-Tetrachlorvinphos∣2,3,5,6-Tetrachloronitrobenzene∣Acrinathrin∣Aldrin∣Ametryn∣Atraton∣Atrazine∣Azinphos-ethyl∣Beflubutamid∣Benalaxyl∣Benfluralin∣Bifenox∣Biphenyl∣Bromophos-ethyl∣Butachlor∣Butamifos∣Carbofuran∣Chlorfenson∣Chlorfenvinphos∣Chlorobenzilate∣Chloroneb∣Chlorpropham∣Chlorpyrifos∣Chlorpyrifos-methyl∣Clomazone∣Coumaphos∣Cyfluthrin∣Cyproconazole∣Cyprodinil∣Desmetryn∣Diazinon∣Diclofop methyl∣Dicrotophos∣Dieldrin∣Difenoconazole∣Diniconazole∣Diphenylamine∣Dipropetryn∣Ethiolate∣Ethion∣Ethofumesate∣Etoxazole∣Etridiazole∣Etrimfos∣Famphur∣Fenbuconazole∣Fenchlorphos∣Fenitrothion∣Fenobucarb∣Fipronil∣Fluazifop-butyl∣Flucythrinate∣Fludioxonil∣Fluorodifen∣Fluquinconazole∣Hexaconazole∣Iprodione∣Isazofos∣Isocarbofos∣Isofenphos-oxon∣Isoprothiolane∣lambda-Cyhalothrin∣Leptophos∣Malaoxon∣Malathion∣Mefenacet∣Methidathion∣Methoprene∣Methoxychlor∣Monolinuron∣Myclobutanil∣Naled∣Napropamide∣Nitrofen∣Omethoate∣Oxadixyl∣Paclobutrazol∣Parathion-methyl∣Pendimethalin∣Pentachloroaniline∣Phosalone∣Phosfolan∣Phosmet∣Phosphamidon∣Pirimiphos-ethyl∣Procymidone∣Profenofos∣Prometryn∣Propanil∣Propiconazole∣Propyzamide∣Prothiophos∣Pyridaphenthion∣Pyrimethanil∣Quintozene∣Simazine∣tau-Fluvalinate∣Terbuthylazine∣Terbutryn∣Tetraconazole∣Thionazin∣Tolclofos-methyl∣Trans-Chlordane (Gamma Isomer)∣Trichloronate
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Ethyl Acetate
Quantity 1.5 mL
Concentration or Composition (by Analyte or Components) (±)-Fenpropathrin 10 μg/ mL∣(EZ)-Tetrachlorvinphos 10 μg/ mL∣2,3,5,6-Tetrachloronitrobenzene 10 μg/ mL∣Acrinathrin 10 μg/ mL∣Aldrin 10 μg/ mL∣Ametryn 10 μg/ mL∣Atraton 10 μg/ mL∣Atrazine 10 μg/ mL∣Azinphos-ethyl 10 μg/ mL∣Beflubutamid 10 μg/ mL∣Benalaxyl 10 μg/ mL∣Benfluralin 10 μg/ mL∣Bifenox 10 μg/ mL∣Biphenyl 10 μg/ mL∣Bromophos-ethyl 10 μg/ mL∣Butachlor 10 μg/ mL∣Butamifos 10 μg/ mL∣Carbofuran 10 μg/ mL∣Chlorfenson 10 μg/ mL∣Chlorfenvinphos 10 μg/ mL∣Chlorobenzilate 10 μg/ mL∣Chloroneb 10 μg/ mL∣Chlorpropham 10 μg/ mL∣Chlorpyrifos 10 μg/ mL∣Chlorpyrifos-methyl 10 μg/ mL∣Clomazone 10 μg/ mL∣Coumaphos 10 μg/ mL∣Cyfluthrin 10 μg/ mL∣Cyproconazole 10 μg/ mL∣Cyprodinil 10 μg/ mL∣Desmetryn 10 μg/ mL∣Diazinon 10 μg/ mL∣Diclofop methyl 10 μg/ mL∣Dicrotophos 10 μg/ mL∣Dieldrin 10 μg/ mL∣Difenoconazole 10 μg/ mL∣Diniconazole 10 μg/ mL∣Diphenylamine 10 μg/ mL∣Dipropetryn 10 μg/ mL∣Ethiolate 10 μg/ mL∣Ethion 10 μg/ mL∣Ethofumesate 10 μg/ mL∣Etoxazole 10 μg/ mL∣Etridiazole 10 μg/ mL∣Etrimfos 10 μg/ mL∣Famphur 10 μg/ mL∣Fenbuconazole 10 μg/ mL∣Fenchlorphos 10 μg/ mL∣Fenitrothion 10 μg/ mL∣Fenobucarb 10 μg/ mL∣Fipronil 10 μg/ mL∣Fluazifop-butyl 10 μg/ mL∣Flucythrinate 10 μg/ mL∣Fludioxonil 10 μg/ mL∣Fluorodifen 10 μg/ mL∣Fluquinconazole 10 μg/ mL∣Hexaconazole 10 μg/ mL∣Iprodione 10 μg/ mL∣Isazofos 10 μg/ mL∣Isocarbofos 10 μg/ mL∣Isofenphos-oxon 10 μg/ mL∣Isoprothiolane 10 μg/ mL∣lambda-Cyhalothrin 10 μg/ mL∣Leptophos 10 μg/ mL∣Malaoxon 10 μg/ mL∣Malathion 10 μg/ mL∣Mefenacet 10 μg/ mL∣Methidathion 10 μg/ mL∣Methoprene 10 μg/ mL∣Methoxychlor 10 μg/ mL∣Monolinuron 10 μg/ mL∣Myclobutanil 10 μg/ mL∣Naled 10 μg/ mL∣Napropamide 10 μg/ mL∣Nitrofen 10 μg/ mL∣Omethoate 10 μg/ mL∣Oxadixyl 10 μg/ mL∣Paclobutrazol 10 μg/ mL∣Parathion-methyl 10 μg/ mL∣Pendimethalin 10 μg/ mL∣Pentachloroaniline 10 μg/ mL∣Phosalone 10 μg/ mL∣Phosfolan 10 μg/ mL∣Phosmet 10 μg/ mL∣Phosphamidon 10 μg/ mL∣Pirimiphos-ethyl 10 μg/ mL∣Procymidone 10 μg/ mL∣Profenofos 10 μg/ mL∣Prometryn 10 μg/ mL∣Propanil 10 μg/ mL∣Propiconazole 10 μg/ mL∣Propyzamide 10 μg/ mL∣Prothiophos 10 μg/ mL∣Pyridaphenthion 10 μg/ mL∣Pyrimethanil 10 μg/ mL∣Quintozene 10 μg/ mL∣Simazine 10 μg/ mL∣tau-Fluvalinate 10 μg/ mL∣Terbuthylazine 10 μg/ mL∣Terbutryn 10 μg/ mL∣Tetraconazole 10 μg/ mL∣Thionazin 10 μg/ mL∣Tolclofos-methyl 10 μg/ mL∣Trans-Chlordane (Gamma Isomer) 10 μg/ mL∣Trichloronate 10 μg/ mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.