missing translation for 'onlineSavingsMsg'
Learn More

GB 23200.121-2021 Pesticide Mixture 3 50 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30314147
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30314147 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30314147 Supplier Dr. Ehrenstorfer Supplier No. DREA50000723AL
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acephate∣Anilofos∣Cadusafos∣Chlorpyrifos∣Chlorpyrifos-methyl∣Coumaphos∣Demeton (O+S)∣Demeton-S Sulfoxide∣Demeton-S-methyl∣Demeton-S-methyl sulfone∣Demeton-S-methyl sulfoxide∣Diazinon∣Dimethoate∣Disulfoton∣Disulfoton-oxon-sulfon∣Disulfoton-sulfone∣Disulfoton-sulfoxide∣Edifenphos∣EPN∣Ethion∣Ethoprophos∣Fensulfothion-sulfone∣Fenthion∣Fenthion-sulfone∣Fenthion-sulfoxide∣Formothion∣Iprobenfos∣Isazofos∣Isocarbofos∣Isofenphos-methyl∣Malaoxon∣Malathion∣Methacrifos∣Methidathion∣Omethoate∣Parathion-ethyl∣Phenthoate∣Phorate∣Phorate-sulfone∣Phorate-sulfoxide∣Phosmet∣Phosmetoxon∣Phoxim∣Pirimiphos-methyl∣Profenofos∣Quinalphos∣Sulfotep∣Terbufos∣Terbufos-sulfone∣Terbufos-sulfoxide∣Thidiazuron∣Tolclofos-methyl∣Triazophos∣Vamidothion
CAS 30560-19-1, 64249-01-0, 95465-99-9, 2921-88-2, 5598-13-0, 56-72-4
Analyte or Component Names Acephate∣Anilofos∣Cadusafos∣Chlorpyrifos∣Chlorpyrifos-methyl∣Coumaphos∣Demeton (O+S)∣Demeton-S Sulfoxide∣Demeton-S-methyl∣Demeton-S-methyl sulfone∣Demeton-S-methyl sulfoxide∣Diazinon∣Dimethoate∣Disulfoton∣Disulfoton-oxon-sulfon∣Disulfoton-sulfone∣Disulfoton-sulfoxide∣Edifenphos∣EPN∣Ethion∣Ethoprophos∣Fensulfothion-sulfone∣Fenthion∣Fenthion-sulfone∣Fenthion-sulfoxide∣Formothion∣Iprobenfos∣Isazofos∣Isocarbofos∣Isofenphos-methyl∣Malaoxon∣Malathion∣Methacrifos∣Methidathion∣Omethoate∣Parathion-ethyl∣Phenthoate∣Phorate∣Phorate-sulfone∣Phorate-sulfoxide∣Phosmet∣Phosmetoxon∣Phoxim∣Pirimiphos-methyl∣Profenofos∣Quinalphos∣Sulfotep∣Terbufos∣Terbufos-sulfone∣Terbufos-sulfoxide∣Thidiazuron∣Tolclofos-methyl∣Triazophos∣Vamidothion
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Solvent or Matrix Acetonitrile
Quantity 1 mL
Concentration or Composition (by Analyte or Components) Acephate 50 μg/mL∣Anilofos 50 μg/mL∣Cadusafos 50 μg/mL∣Chlorpyrifos 50 μg/mL∣Chlorpyrifos-methyl 50 μg/mL∣Coumaphos 50 μg/mL∣Demeton (O+S) 50 μg/mL∣Demeton-S Sulfoxide 50 μg/mL∣Demeton-S-methyl 50 μg/mL∣Demeton-S-methyl sulfone 50 μg/mL∣Demeton-S-methyl sulfoxide 50 μg/mL∣Diazinon 50 μg/mL∣Dimethoate 50 μg/mL∣Disulfoton 50 μg/mL∣Disulfoton-oxon-sulfon 50 μg/mL∣Disulfoton-sulfone 50 μg/mL∣Disulfoton-sulfoxide 50 μg/mL∣Edifenphos 50 μg/mL∣EPN 50 μg/mL∣Ethion 50 μg/mL∣Ethoprophos 50 μg/mL∣Fensulfothion-sulfone 50 μg/mL∣Fenthion 50 μg/mL∣Fenthion-sulfone 50 μg/mL∣Fenthion-sulfoxide 50 μg/mL∣Formothion 50 μg/mL∣Iprobenfos 50 μg/mL∣Isazofos 50 μg/mL∣Isocarbofos 50 μg/mL∣Isofenphos-methyl 50 μg/mL∣Malaoxon 50 μg/mL∣Malathion 50 μg/mL∣Methacrifos 50 μg/mL∣Methidathion 50 μg/mL∣Omethoate 50 μg/mL∣Parathion-ethyl 50 μg/mL∣Phenthoate 50 μg/mL∣Phorate 50 μg/mL∣Phorate-sulfone 50 μg/mL∣Phorate-sulfoxide 50 μg/mL∣Phosmet 50 μg/mL∣Phosmetoxon 50 μg/mL∣Phoxim 50 μg/mL∣Pirimiphos-methyl 50 μg/mL∣Profenofos 50 μg/mL∣Quinalphos 50 μg/mL∣Sulfotep 50 μg/mL∣Terbufos 50 μg/mL∣Terbufos-sulfone 50 μg/mL∣Terbufos-sulfoxide 50 μg/mL∣Thidiazuron 50 μg/mL∣Tolclofos-methyl 50 μg/mL∣Triazophos 50 μg/mL∣Vamidothion 50 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.