missing translation for 'onlineSavingsMsg'
Learn More

GB 23200.121-2021 Pesticide Mixture 4 50 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30316944
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30316944 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30316944 Supplier Dr. Ehrenstorfer Supplier No. DREA50000724AL
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifica

Chemical Name or Material Chlorfluazuron∣Chlorobenzuron∣Fenamiphos∣Fenamiphos-sulfone∣Fenamiphos-sulfoxide∣Hexaflumuron∣Imazalil∣Novaluron∣Phosfolan∣Phospholan-methyl∣Procymidone∣Pyribenzoxim∣Teflubenzuron∣Triflumuron
CAS 71422-67-8, 57160-47-1, 22224-92-6, 31972-44-8, 31972-43-7, 86479-06-3
Analyte or Component Names Chlorfluazuron∣Chlorobenzuron∣Fenamiphos∣Fenamiphos-sulfone∣Fenamiphos-sulfoxide∣Hexaflumuron∣Imazalil∣Novaluron∣Phosfolan∣Phospholan-methyl∣Procymidone∣Pyribenzoxim∣Teflubenzuron∣Triflumuron
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Solvent or Matrix Acetonitrile
Quantity 1 mL
Concentration or Composition (by Analyte or Components) Chlorfluazuron 50 μg/mL∣Chlorobenzuron 50 μg/mL∣Fenamiphos 50 μg/mL∣Fenamiphos-sulfone 50 μg/mL∣Fenamiphos-sulfoxide 50 μg/mL∣Hexaflumuron 50 μg/mL∣Imazalil 50 μg/mL∣Novaluron 50 μg/mL∣Phosfolan 50 μg/mL∣Phospholan-methyl 50 μg/mL∣Procymidone 50 μg/mL∣Pyribenzoxim 50 μg/mL∣Teflubenzuron 50 μg/mL∣Triflumuron 50 μg/mL
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.