missing translation for 'onlineSavingsMsg'
Learn More

GB 23200.121-2021 Pesticide Mixture 5 50 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30311132
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30311132 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30311132 Supplier Dr. Ehrenstorfer Supplier No. DREA50000725AL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acetochlor∣Alachlor∣Amisulbrom∣Azinphos-methyl∣Benalaxyl∣Benzovindiflupyr∣Benzoximate∣Boscalid∣Butachlor∣Carboxin∣Chloridazon∣Chromafenozide∣Clethodim Sulfone∣Clomazone∣Cyantraniliprole∣Cyazofamid∣Cyazofamid-dessulfonamide∣Cyflufenamid∣Dichlorvos∣Dicrotophos∣Diflufenican∣Dimethenamid∣Dimethomorph∣Dimoxystrobin∣Ethiprole∣Ethirimol∣Fenamidone∣Fenaminostrobin∣Fenhexamid∣Fenoxanil∣Fensulfothion∣Fensulfothion-oxon∣Fensulfothion-oxon-sulfone∣Fipronil∣Fipronil Sulfone∣Flonicamid∣Florasulam∣Flubendiamide∣Flufenacet∣Flumetsulam∣Flumorph∣Fluopicolide∣Fluopyram
CAS 34256-82-1, 15972-60-8, 348635-87-0, 86-50-0, 71626-11-4, 1072957-71-1
Analyte or Component Names Acetochlor∣Alachlor∣Amisulbrom∣Azinphos-methyl∣Benalaxyl∣Benzovindiflupyr∣Benzoximate∣Boscalid∣Butachlor∣Carboxin∣Chloridazon∣Chromafenozide∣Clethodim Sulfone∣Clomazone∣Cyantraniliprole∣Cyazofamid∣Cyazofamid-dessulfonamide∣Cyflufenamid∣Dichlorvos∣Dicrotophos∣Diflufenican∣Dimethenamid∣Dimethomorph∣Dimoxystrobin∣Ethiprole∣Ethirimol∣Fenamidone∣Fenaminostrobin∣Fenhexamid∣Fenoxanil∣Fensulfothion∣Fensulfothion-oxon∣Fensulfothion-oxon-sulfone∣Fipronil∣Fipronil Sulfone∣Flonicamid∣Florasulam∣Flubendiamide∣Flufenacet∣Flumetsulam∣Flumorph∣Fluopicolide∣Fluopyram
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Solvent or Matrix Acetonitrile
Quantity 1 mL
Concentration or Composition (by Analyte or Components) Acetochlor 50 μg/mL∣Alachlor 50 μg/mL∣Amisulbrom 50 μg/mL∣Azinphos-methyl 50 μg/mL∣Benalaxyl 50 μg/mL∣Benzovindiflupyr 50 μg/mL∣Benzoximate 50 μg/mL∣Boscalid 50 μg/mL∣Butachlor 50 μg/mL∣Carboxin 50 μg/mL∣Chloridazon 50 μg/mL∣Chromafenozide 50 μg/mL∣Clethodim Sulfone 50 μg/mL∣Clomazone 50 μg/mL∣Cyantraniliprole 50 μg/mL∣Cyazofamid 50 μg/mL∣Cyazofamid-dessulfonamide 50 μg/mL∣Cyflufenamid 50 μg/mL∣Dichlorvos 50 μg/mL∣Dicrotophos 50 μg/mL∣Diflufenican 50 μg/mL∣Dimethenamid 50 μg/mL∣Dimethomorph 50 μg/mL∣Dimoxystrobin 50 μg/mL∣Ethiprole 50 μg/mL∣Ethirimol 50 μg/mL∣Fenamidone 50 μg/mL∣Fenaminostrobin 50 μg/mL∣Fenhexamid 50 μg/mL∣Fenoxanil 50 μg/mL∣Fensulfothion 50 μg/mL∣Fensulfothion-oxon 50 μg/mL∣Fensulfothion-oxon-sulfone 50 μg/mL∣Fipronil 50 μg/mL∣Fipronil Sulfone 50 μg/mL∣Flonicamid 50 μg/mL∣Florasulam 50 μg/mL∣Flubendiamide 50 μg/mL∣Flufenacet 50 μg/mL∣Flumetsulam 50 μg/mL∣Flumorph 50 μg/mL∣Fluopicolide 50 μg/mL∣Fluopyram 50 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.