missing translation for 'onlineSavingsMsg'
Learn More

GB 23200.121-2021 Pesticide Mixture 6 50 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30315521
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30315521 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30315521 Supplier Dr. Ehrenstorfer Supplier No. DREA50000726AL
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (±)-Metamifop∣Fluthiacet-methyl∣Flutolanil∣Fluxapyroxad∣Fonofos∣Heptenophos∣Hexazinone∣Iprodione∣Isopyrazam∣Isoxaflutole∣Mandipropamid∣Mefenacet∣Mepronil∣Metalaxyl∣Metamitron∣Methamidophos∣Methoxyfenozide∣Mevinphos∣Monocrotophos∣Napropamide∣Oxamyl-oxime∣Oxaziclomefone∣Penflufen∣Penoxsulam∣Penthiopyrad∣Phosphamidon∣Picolinafen∣Pretilachlor∣Probenazole∣Propachlor∣Propanil∣Propisochlor∣Propyzamide∣Proquinazid∣Pyridaben∣Pyrimorph∣Saflufenacil∣Sedaxane∣Silthiofarm∣S-Metolachlor∣Spirotetramat∣Spirotetramat-enol-glucoside∣Spirotetramat-keto-hydroxy∣Spirotetramat-mono-hydroxy∣Sulfoxaflor∣Tebufenozide∣Tolfenpyrad∣Zoxamide
CAS 256412-89-2, 117337-19-6, 66332-96-5, 907204-31-3, 944-22-9, 23560-59-0
Analyte or Component Names (±)-Metamifop∣Fluthiacet-methyl∣Flutolanil∣Fluxapyroxad∣Fonofos∣Heptenophos∣Hexazinone∣Iprodione∣Isopyrazam∣Isoxaflutole∣Mandipropamid∣Mefenacet∣Mepronil∣Metalaxyl∣Metamitron∣Methamidophos∣Methoxyfenozide∣Mevinphos∣Monocrotophos∣Napropamide∣Oxamyl-oxime∣Oxaziclomefone∣Penflufen∣Penoxsulam∣Penthiopyrad∣Phosphamidon∣Picolinafen∣Pretilachlor∣Probenazole∣Propachlor∣Propanil∣Propisochlor∣Propyzamide∣Proquinazid∣Pyridaben∣Pyrimorph∣Saflufenacil∣Sedaxane∣Silthiofarm∣S-Metolachlor∣Spirotetramat∣Spirotetramat-enol-glucoside∣Spirotetramat-keto-hydroxy∣Spirotetramat-mono-hydroxy∣Sulfoxaflor∣Tebufenozide∣Tolfenpyrad∣Zoxamide
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Solvent or Matrix Acetonitrile
Quantity 1 mL
Concentration or Composition (by Analyte or Components) (±)-Metamifop 50 μg/mL∣Fluthiacet-methyl 50 μg/mL∣Flutolanil 50 μg/mL∣Fluxapyroxad 50 μg/mL∣Fonofos 50 μg/mL∣Heptenophos 50 μg/mL∣Hexazinone 50 μg/mL∣Iprodione 50 μg/mL∣Isopyrazam 50 μg/mL∣Isoxaflutole 50 μg/mL∣Mandipropamid 50 μg/mL∣Mefenacet 50 μg/mL∣Mepronil 50 μg/mL∣Metalaxyl 50 μg/mL∣Metamitron 50 μg/mL∣Methamidophos 50 μg/mL∣Methoxyfenozide 50 μg/mL∣Mevinphos 50 μg/mL∣Monocrotophos 50 μg/mL∣Napropamide 50 μg/mL∣Oxamyl-oxime 50 μg/mL∣Oxaziclomefone 50 μg/mL∣Penflufen 50 μg/mL∣Penoxsulam 50 μg/mL∣Penthiopyrad 50 μg/mL∣Phosphamidon 50 μg/mL∣Picolinafen 50 μg/mL∣Pretilachlor 50 μg/mL∣Probenazole 50 μg/mL∣Propachlor 50 μg/mL∣Propanil 50 μg/mL∣Propisochlor 50 μg/mL∣Propyzamide 50 μg/mL∣Proquinazid 50 μg/mL∣Pyridaben 50 μg/mL∣Pyrimorph 50 μg/mL∣Saflufenacil 50 μg/mL∣Sedaxane 50 μg/mL∣Silthiofarm 50 μg/mL∣S-Metolachlor 50 μg/mL∣Spirotetramat 50 μg/mL∣Spirotetramat-enol-glucoside 50 μg/mL∣Spirotetramat-keto-hydroxy 50 μg/mL∣Spirotetramat-mono-hydroxy 50 μg/mL∣Sulfoxaflor 50 μg/mL∣Tebufenozide 50 μg/mL∣Tolfenpyrad 50 μg/mL∣Zoxamide 50 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.