missing translation for 'onlineSavingsMsg'
Learn More

GB/T 21318-2007, SN/T 1928-2007 10 Nitroimidazoles 100 μg/mL in Methanol, Dr. Ehrenstorfer

Product Code. 30320268
Change view
Click to view available options
Quantity:
1.5 mL
Unit Size:
1.5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30320268 1.5 mL 1.5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30320268 Supplier Dr. Ehrenstorfer Supplier No. DREA50000088ME

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Methyl-4-nitroimidazole∣4-Nitroimidazole∣5-Chloro-1-Methyl-4-Nitroimidazole∣5-Nitrobenzimidazole∣Dimetridazole∣Dimetridazole-2-hydroxy∣Ipronidazole∣Metronidazole∣Metronidazole-hydroxy∣Ronidazole (RNZ)
CAS 696-23-1, 3034-38-6, 4897-25-0, 94-52-0, 551-92-8, 936-05-0
Analyte or Component Names 2-Methyl-4-nitroimidazole∣4-Nitroimidazole∣5-Chloro-1-Methyl-4-Nitroimidazole∣5-Nitrobenzimidazole∣Dimetridazole∣Dimetridazole-2-hydroxy∣Ipronidazole∣Metronidazole∣Metronidazole-hydroxy∣Ronidazole (RNZ)
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Methanol
Quantity 1.5 mL
Concentration or Composition (by Analyte or Components) 2-Methyl-4-nitroimidazole 100 μg/mL∣4-Nitroimidazole 100 μg/mL∣5-Chloro-1-Methyl-4-Nitroimidazole 100 μg/mL∣5-Nitrobenzimidazole 100 μg/mL∣Dimetridazole 100 μg/mL∣Dimetridazole-2-hydroxy 100 μg/mL∣Ipronidazole 100 μg/mL∣Metronidazole 100 μg/mL∣Metronidazole-hydroxy 100 μg/mL∣Ronidazole (RNZ) 100 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.