missing translation for 'onlineSavingsMsg'
Learn More

GB/T 22286-2008 11 β-Agonists 100 μg/mL in Methanol, Dr. Ehrenstorfer

Product Code. 30312383
Change view
Click to view available options
Quantity:
1.5 mL
Unit Size:
1.5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30312383 1.5 mL 1.5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30312383 Supplier Dr. Ehrenstorfer Supplier No. DREA50000096ME
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Brombuterol hydrochloride∣Bromchlorbuterol hydrochloride∣Cimaterol∣Cimbuterol∣Clenbuterol hydrochloride∣Isoxsuprine hydrochloride∣Mabuterol∣Mapenterol hydrochloride∣Ractopamine hydrochloride∣Salbutamol sulfate∣Terbutaline
CAS 21912-49-2, 78982-84-0, 54239-37-1, 54239-39-3, 21898-19-1, 579-56-6
Analyte or Component Names Brombuterol hydrochloride∣Bromchlorbuterol hydrochloride∣Cimaterol∣Cimbuterol∣Clenbuterol hydrochloride∣Isoxsuprine hydrochloride∣Mabuterol∣Mapenterol hydrochloride∣Ractopamine hydrochloride∣Salbutamol sulfate∣Terbutaline
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Methanol
Quantity 1.5 mL
Concentration or Composition (by Analyte or Components) Brombuterol hydrochloride 100 μg/mL∣Bromchlorbuterol hydrochloride 100 μg/mL∣Cimaterol 100 μg/mL∣Cimbuterol 100 μg/mL∣Clenbuterol hydrochloride 100 μg/mL∣Isoxsuprine hydrochloride 100 μg/mL∣Mabuterol 100 μg/mL∣Mapenterol hydrochloride 100 μg/mL∣Ractopamine hydrochloride 100 μg/mL∣Salbutamol sulfate 100 μg/mL∣Terbutaline 100 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.