missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GB/T 39665-2020 Pesticide Mixture 606 100 μg/mL in Methanol, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Carbaryl∣Carbendazim∣Diallate∣Dinoterb∣Diuron∣Epoxiconazole∣Fenpropimorph∣Fluazifop-P-butyl∣Flumioxazin∣Ioxynil∣Iprodione∣Isoxaflutole∣Kresoxim-methyl∣Linuron∣Molinate∣Monocrotophos∣Propargite∣Propazine∣Simazine∣Tridemorph |
| CAS | 63-25-2, 10605-21-7, 2303-16-4, 1420-07-1, 330-54-1, 133855-98-8 |
| Analyte or Component Names | Carbaryl∣Carbendazim∣Diallate∣Dinoterb∣Diuron∣Epoxiconazole∣Fenpropimorph∣Fluazifop-P-butyl∣Flumioxazin∣Ioxynil∣Iprodione∣Isoxaflutole∣Kresoxim-methyl∣Linuron∣Molinate∣Monocrotophos∣Propargite∣Propazine∣Simazine∣Tridemorph |
| Shipping Condition | +20°C |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Methanol |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | Carbaryl 100 μg/mL∣Carbendazim 100 μg/mL∣Diallate 100 μg/mL∣Dinoterb 100 μg/mL∣Diuron 100 μg/mL∣Epoxiconazole 100 μg/mL∣Fenpropimorph 100 μg/mL∣Fluazifop-P-butyl 100 μg/mL∣Flumioxazin 100 μg/mL∣Ioxynil 100 μg/mL∣Iprodione 100 μg/mL∣Isoxaflutole 100 μg/mL∣Kresoxim-methyl 100 μg/mL∣Linuron 100 μg/mL∣Molinate 100 μg/mL∣Monocrotophos 100 μg/mL∣Propargite 100 μg/mL∣Propazine 100 μg/mL∣Simazine 100 μg/mL∣Tridemorph 100 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?