missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GBAS Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-16616
This item is not returnable.
View return policy
Description
GBAS Polyclonal specifically detects GBAS in Human samples. It is validated for Western Blot.
Specifications
| GBAS | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| glioblastoma amplified sequence, Glioblastoma-amplified sequence, NIPSNAP2NipSnap2,4-nitrophenylphosphatase domain and non-neuronal SNAP25-like 2, protein NipSnap homolog 2 | |
| Rabbit | |
| 34 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG |
| Western Blot | |
| 0.89mg/mL | |
| Western Blot 1:500-1:3000 | |
| O75323 | |
| NIPSNAP2 | |
| Recombinant protein encompassing a sequence within the center region of human GBAS. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 2631 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction