missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GBP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-03972
This item is not returnable.
View return policy
Description
GBP1 Polyclonal specifically detects GBP1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| GBP1 | |
| Polyclonal | |
| Western Blot 2-4 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 5 ug/ml | |
| NP_002044.2 | |
| GBP1 | |
| A portion of amino acids 500-550 of human GBP1 was used as the immunogen for this antibody. | |
| 0.1 mg | |
| Primary | |
| Reacts with Human and Mouse. The amino acid sequence used as immunogen has 100% identity in human, 95% in rat and 79% in mouse. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| GBP-1, GTP-binding protein 1, Guanine nucleotide-binding protein 1, guanylate binding protein 1, interferon-inducible, guanylate binding protein 1, interferon-inducible, 67kDa, huGBP-1, interferon-induced guanylate-binding protein 1 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 2633 | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction