missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GBP4 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15836063
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15836063 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15836063 Supplier Invitrogen™ Supplier No. PA567644

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Predicted to react with Mouse and Rat samples.

Guanylate-binding proteins, such as GBP4, are induced by interferon and hydrolyze GTP to both GDP and GMP.[supplied by OMIM, Dec 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen GBP4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GBP4
Gene Accession No. A4UUI3, Q96PP9
Gene Alias AW228052; Gbp3; GBP4; GBP-4; GTP-binding protein 4; guanine nucleotide-binding protein 4; guanylate binding protein 4; guanylate nucleotide binding protein 3; guanylate nucleotide binding protein 4; guanylate-binding protein 4; guanylate-binding protein 4 homolog; macrophage activation 2; Mag-2; mKIAA4245; Mpa2; Mpa-2
Gene Symbols GBP4
Host Species Rabbit
Immunogen Synthesized peptide derived from human GBP4.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115361, 229900, 310917
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.