missing translation for 'onlineSavingsMsg'
Learn More

GBR 12909 dihydrochloride, Thermo Scientific Chemicals

Product Code. 15423419
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15423419 10 mg 10mg
15433419 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15423419 Supplier Thermo Scientific Alfa Aesar Supplier No. J60684.MA
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
This item is not returnable. View return policy

Dopamine uptake inhibitor

Dopamine uptake inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67469-78-7
Molecular Formula C28H34Cl2F2N2O
Molecular Weight (g/mol) 523.49
MDL Number MFCD00055193
InChI Key MIBSKSYCRFWIRU-UHFFFAOYSA-N
PubChem CID 104920
ChEBI CHEBI:64086
IUPAC Name 1-[2-[bis(4-fluorophenyl)methoxy]ethyl]-4-(3-phenylpropyl)piperazine;dihydrochloride
SMILES C1CN(CCN1CCCC2=CC=CC=C2)CCOC(C3=CC=C(C=C3)F)C4=CC=C(C=C4)F.Cl.Cl

Specifications

Color White
Quantity 10 mg
Synonym vanoxerine dihydrochloride, gbr 12909 dihydrochloride, vanoxeamine, gbr-12909 dihydrochloride, vanoxerine hydrochloride, unii-mwo1ip03ev, mwo1ip03ev, vanoxerine dihydrochloride gbr12909, gbr12909 dihydrochloride, 1-2-bis 4-fluorophenyl methoxy ethyl-4-3-phenylpropyl piperazine dihydrochloride
Solubility Information Soluble in DMSO to 50mM; Soluble to 5mM in water with warming.
Formula Weight 523.49
Physical Form Powder
Chemical Name or Material GBR 12909 dihydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.