missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GCH1 Monoclonal Antibody (OTI2D12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810248
This item is not returnable.
View return policy
Description
This gene encodes a member of the GTP cyclohydrolase family. The encoded protein is the first and rate-limiting enzyme in tetrahydrobiopterin biosynthesis, catalyzing the conversion of GTP into 7,8-dihydroneopterin triphosphate. BH4 is an essential cofactor required by aromatic amino acid hydroxylases as well as nitric oxide synthases. Mutations in this gene are associated with maligt hyperphenylalaninemia and dopa-responsive dystonia. Several alternatively spliced transcript variants encoding different isoforms have been described; however, not all variants give rise to a functional enzyme.Specifications
| GCH1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P30793 | |
| GCH1 | |
| Full length human recombit protein of human GCH1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2D12 | |
| Unconjugated | |
| GCH1 | |
| DYT14, DYT5, DYT5a, GCH, GTP-CH-1, GTPCH1, HPABH4B | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 2643 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction