Learn More
Invitrogen™ GCLC Recombinant Rabbit Monoclonal Antibody (24GB4860)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557408
 View more versions of this product
Product Code. 30275768
  • 564.00 € / 100µL
Description
GCLC Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Glutamate-cysteine ligase, also known as gamma-glutamylcysteine synthetase is the first rate-limiting enzyme of glutathione synthesis. The enzyme consists of two subunits, a heavy catalytic subunit and a light regulatory subunit. This locus encodes the catalytic subunit, while the regulatory subunit is derived from a different gene located on chromosome 1p22-p21. Mutations at this locus have been associated with hemolytic anemia due to deficiency of gamma-glutamylcysteine synthetase and susceptibility to myocardial infarction.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GCLC
Recombinant Monoclonal
Unconjugated
Gclc
cb1049; D9Wsu168e; EGK_15023; fe36e11; gamma GCS-HS; gamma-ECS; Gamma-glutamylcysteine synthetase; gamma-glutamylcysteine synthetase heavy subunit; GCL; Gclc; GCS; GCS heavy chain; Ggcs-hs; GLCL; GLCLC; GLCL-H; glutamate-cysteine ligase catalytic subunit; Glutamate--cysteine ligase catalytic subunit; glutamate-cysteine ligase, catalytic subunit; Glutamylcysteine gamma synthetase light chain; wu:fe36e11
Rabbit
Affinity chromatography
RUO
14629, 25283, 2729
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB4860
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P19468, P48506, P97494
Gclc
Recombinant protein of human GCLC (aa 1-300).
100 μL
Primary
Human, Mouse, Rat