missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GCSAM Recombinant Rabbit Monoclonal Antibody (K01_1R55)

Product Code. p-200114856
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30830099 100 μL
30830434 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30830099 Supplier Invitrogen™ Supplier No. MA563602

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: MGNSLLRENR RQQNTQEMPW NVRMQSPKQR TSRCWDHHIA EGCFCLPWKK ILIFEKRQDS QNENERMSST PIQDNVDQTY SEELCYTLIN HRVLCTRPSG NSAEEYYENV PCKAERPRES LGGTETEYSL LHMPSTDPRH ARSPEDEYEL LMPHRISSHF LQQPRPLMAP SETQFSHL Highest antigen sequence identity to the following orthologs: Mouse - 55%, Pig - 67%, Cynomolgus monkey - 92%.

GCET2 is a protein which may function in signal transduction pathways and whose expression is elevated in germinal cell lymphomas. It contains a putative PDZ-interacting domain, an immunoreceptor tyrosine-based activation motif (ITAM), and two putative SH2 binding sites. In B cells, its expression is specifically induced by interleukin-4.This gene encodes a protein which may function in signal transduction pathways and whose expression is elevated in germinal cell lymphomas. It contains a putative PDZ-interacting domain, an immunoreceptor tyrosine-based activation motif (ITAM), and two putative SH2 binding sites. In B cells, its expression is specifically induced by interleukin-4. Alternative splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GCSAM
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_1R55
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 40% glycerol, 0.05% BSA, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene GCSAM
Gene Accession No. Q8N6F7
Gene Alias GAL; GCAT2; Gcet; GCET2; GCET2 germinal center expressed transcript 2; GCSAM; Germinal center associated lymphoma (hGAL); germinal center associated signaling and motility; germinal center associated, signaling and motility; germinal center B-cell-expressed transcript 2 protein; germinal center expressed transcript 2; germinal center-associated lymphoma protein; Germinal center-associated signaling and motility protein; germinal center-associated, signaling and motility; HGAL; human germinal center-associated lymphoma; M17; M17-L; MGC40441
Gene Symbols GCSAM
Host Species Rabbit
Immunogen Recombinant protein of human GCET2 (aa 1-178).
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 257144
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.