Learn More
GDF-15 Antibody (CL0328) [DyLight 650], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-52917C
Product Code. 30302845
  • 737.00 € / 0.10mL
Description
GDF-15 Monoclonal antibody specifically detects GDF-15 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GDF-15
Monoclonal
DyLight 650
growth differentiation factor 15, growth/differentiation factor 15, Macrophage inhibitory cytokine 1, MIC-1NSAID-activated gene 1 protein, MIC1Prostate differentiation factor, NAG-1NSAID-regulated gene 1 protein, NSAID (nonsteroidal inflammatory drug)-activated protein 1, PDFGDF-15, PLABNRG-1, Placental bone morphogenetic protein, Placental TGF-beta, PTGFBPTGF-beta
This antibody was developed using a recombinant protein derived from Q99988, with the exact immunogen sequence remaining proprietary.
0.1 mL
Hepatic Stem Cell Markers, Stem Cell Markers
9518
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown, Immunocytochemistry/Immunofluorescence
CL0328
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified