missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GDF10 Polyclonal Antibody
GREENER_CHOICE

Product Code. 30535585
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30535585 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30535585 Supplier Invitrogen™ Supplier No. PA5148504

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

Reconstitute at 0.2 mg/mL in sterile PBS.

The protein encoded by this gene is a member of the bone morphogenetic protein (BMP) family and the TGF-beta superfamily. This group of proteins is characterized by a polybasic proteolytic processing site which is cleaved to produce a mature protein containing seven conserved cysteine residues. The members of this family are regulators of cell growth and differentiation in both embryonic and adult tissues. Studies in mice suggest that the protein encoded by this gene plays a role in skeletal morphogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GDF10
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with trehalose and no preservative
Gene GDF10
Gene Accession No. P55107
Gene Alias BIP; Bmp3b; BMP-3B; Bone morphogenetic protein 3B; bone morphogenetic protein 3B; growth/differentiation factor 10; bone-inducing protein; Gdf10; GDF-10; growth differentiation factor 10; growth/differentiation factor 10; prepro bone inducing protein
Gene Symbols GDF10
Host Species Goat
Immunogen E. coli-derived recombinant human BMP-3b/GDF-10, Gln369-Arg478.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2662
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.