missing translation for 'onlineSavingsMsg'
Learn More

PeproTech GDF3 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Product Code. 17855793
Change view
Click to view available options
Quantity:
2 x 500 μg
25 μg
50 μg
500 μg
Unit Size:
1mg
25µg
500µg
50µg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17855793 50 μg 50µg
17895813 25 μg 25µg
17845793 2 x 500 μg 1mg
30054693 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 17855793 Supplier PeproTech Supplier No. 500P235BT50UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

AA Sequence of recombinant protein: AAIPVPKLSC KNLCHRHQLF INFRDLGWHK WIIAPKGFMA NYCHGECPFS LTISLNSSNY AFMQALMHAV DPEIPQAVCI PTKLSPISML YQDNNDNVIL RHYEDMVVDE CGCG. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human GDF-3. Anti-Human GDF-3-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect hGDF-3 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human GDF-3 (500-P235) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant hGDF-3. Western Blot: To detect hGDF-3 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hGDF-3 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P235BT-1MG will be provided as 2 x 500 μg

GDF3 is a member of the bone morphogenetic protein (BMP) family and the TGF-beta superfamily. This group of proteins is characterized by a polybasic proteolytic processing site which is cleaved to produce a mature protein containing seven conserved cysteine residues. The members of this family are regulators of cell growth and differentiation in both embryonic and adult tissues. The function of this protein is unknown, but expression studies suggest it may be involved in regulation of the adult lymphatic and erythroid systems and embryonic development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GDF3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.1-1.0 mg/mL
Conjugate Biotin
Formulation PBS with no preservative
Gene GDF3
Gene Accession No. Q9NR23
Gene Alias C78318; ecat9; Gdf3; Gdf-3; growth differentiation factor 3; growth/differentiation factor 3; KFS3; MCOP7; MCOPCB6; RGD1564178; UNQ222/PRO248; VG-1-related protein 2; Vgr2; Vgr-2
Gene Symbols GDF3
Host Species Rabbit
Immunogen E.coli-derived Recombinant Human GDF-3.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9573
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.