missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GDF9 Monoclonal Antibody (785123)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA561811
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
GDF9 is a member of the bone morphogenetic protein (BMP) family and the TGF-beta superfamily. This group of proteins is characterized by a polybasic proteolytic processing site which is cleaved to produce a mature protein containing seven conserved cysteine residues. The members of this family are regulators of cell growth and differentiation in both embryonic and adult tissues.Growth factors synthesized by ovarian somatic cells directly affect oocyte growth and function. GDF9 is expressed in oocytes and is thought to be required for ovarian folliculogenesis.
Specifications
| GDF9 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q07105 | |
| GDF9 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse GDF-9, Met1-Arg441. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a |
| Neutralization | |
| 785123 | |
| Unconjugated | |
| GDF9 | |
| Gdf9; Gdf-9; growth differentiation factor 9; growth/differentiation factor 9 | |
| Rat | |
| Protein A/G | |
| RUO | |
| 14566 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction