missing translation for 'onlineSavingsMsg'
Learn More

Melford Geldanamycin

Product Code. 30018302
Change view
Click to view available options
Quantity:
1 mg
5 mg
500 μg
Unit Size:
1mg
500µg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018302 500 μg 500µg
30018304 5 mg 5mg
30018303 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30018302 Supplier Melford Supplier No. G400400.0005
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Geldanamycin is a naturally occurring antibiotic derived from the bacterium Streptomyces hygroscopicus.

Geldanamycin is a naturally occurring antibiotic derived from the bacterium Streptomyces hygroscopicus. It has gained significance due to its role as an inhibitor of heat shock protein 90 (Hsp90), a molecular chaperone involved in protein folding and stabilization.

Geldanamycin is often used as a research tool to study cellular processes that are dependent on Hsp90. By inhibiting Hsp90, geldanamycin affects the stability and function of various client proteins associated with cell signaling, growth, and survival.

Researchers use geldanamycin to investigate the role of Hsp90 in the context of cancer, as many oncoproteins require Hsp90 for proper folding and stability.

Specifications

Chemical Name or Material Geldanamycin
CAS 30562-34-6
Quantity 500 μg
Molecular Formula C29H40N2O9
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.