missing translation for 'onlineSavingsMsg'
Learn More

Gemfibrozil, MedChemExpress

Product Code. 16448196
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16448196 100 mg 100mg
16422076 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16448196 Supplier Medchem Express Supplier No. HYB0258
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Gemfibrozil is an activator of PPAR-α, used as a lipid-lowering drug; Gemfibrozil is also a nonselective inhibitor of several P450 isoforms, with Ki values for CYP2C9, 2C19, 2C8, and 1A2 of 5.8, 24, 69, and 82 μM, respectively.

Gemfibrozil is an activator of PPAR-α, used as a lipid-lowering drug

Specifications

Chemical Name or Material Gemfibrozil
CAS 25812-30-0
Color White
Purity Grade Notes Research
Molecular Formula C15H22O3
Quantity 100 mg
Synonym CI-719
Solubility Information DMSO : 100 mg/mL (399.47 mM; Need ultrasonic)
SMILES O=C(O)C(C)(C)CCCOC1=CC(C)=CC=C1C
Molecular Weight (g/mol) 250.33
Formula Weight 250.33
Percent Purity 99.62%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.