missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Gemtuzumab Recombinant Monoclonal Antibody

Recombinant Monoclonal Antibody
477.00 € - 1204.00 €
Specifications
| Antigen | Gemtuzumab Humanized |
|---|---|
| Concentration | 1 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30284735
![]() |
Invitrogen™
MA560069 |
100 μg |
477.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30283856
|
Invitrogen™
MA560070 |
1 mg |
1204.00 €
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Gemtuzumab ozogamicin is a recombinant humanized IgG4 kappa antibody which is conjugated with calicheamicin derivative, a cytotoxic antitumor antibiotic isolated from fermentation of Micromonospora echinospora ssp. calichensis. Gemtuzumab ozogamicin has approximately 50% of the antibody loaded with 4-6 moles calicheamicin per mole of antibodyLabel. The antibody is specifically directed against the CD33 antigen present on leukemic myeloblasts in most patients with acute myeloid leukemia (AML). By binding to the CD33 antigen on tumors, the cytotoxic agent blocks the growth of cancerous cells and causes cell death.Specifications
| Gemtuzumab Humanized | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Lyophilized | |
| IgG1 κ | |
| Human | |
| CDP-771; CMA-676 (ADC); gemtuzumab ozogamicin; hP67.6; hP67.6-calicheamicin (ADC); WAY-CMA-676 | |
| Antibody |
| 1 mg/mL | |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| Unconjugated | |
| Human | |
| RUO | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Primary | |
| Protein A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title