Learn More
General Purpose Viscosity Standards, Paragon Scientific™
Certified in strict accordance with ASTM D2162, the primary method for viscosity standards manufacture. Dual certified to the International Standards BS EN ISO / IEC 17025 and ISO 17034 under our UKAS accrediatation.
Brand: Paragon Scientific N1.0
Additional Details : CAS Number : 64742-88-7 Weight : 0.40000kg Transport : UN number : 1300 Chem class : 3 Pack group : III
Description
- Certified according to ISO 17025 / ISO 17034 under our UKAS accreditation
- Certified in strict accordance with ASTM D2162
- Density g/mL in accordance with ASTM D1480
- For use with glass capillary viscometers, automatic viscometers and other viscosity measuring equipment
- Kinematic (cSt) & dynamic viscosity (mPa·s) given at all temperatures
- Standard Pack Size 500mL
- Fully traceable to National Standards
- Ensures full compliance to ASTM & IP test method protocol
- Tamper evident security packaging
- 2 Year shelf life
- Manufactured in the United Kingdom
Specifications
500 mL | |
0.9525 mPa.s (25 °C) | |
Certified according to ISO 17025 / ISO 17034 under our UKAS accreditation |
Safety and Handling
- N1.0
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 3
- LONG-TERM AQUATIC HAZARD Chronic 2
- Aspiration toxicity Category 1
- Specific target organ toxicity Category 3
- Specific target organ toxicity Category 1
Hazard Statement
- H226-Flammable liquid and vapour.
- H372-Causes damage to organs.
- H304-May be fatal if swallowed and enters airways.
- H336-May cause drowsiness or dizziness.
- H411-Toxic to aquatic life with long lasting effects.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P273-Avoid release to the environment.
Nominal values quoted