missing translation for 'onlineSavingsMsg'
Learn More

General Purpose Viscosity Standards, Paragon Scientific™

Product Code. 16010272
Change view
Click to view available options
Viscosity:
0.2950 mPa.s (25°C)
0.5204 mPa.s (25°C)
0.9525 mPa.s (25 °C)
100.7 mPa.s (25°C)
10030 mPa.s (25°C)
124.9 mPa.s (25 °C)
1273 mPa.s (25°C)
14.42 mPa.s (25°C)
19588 mPa.s (25°C)
2.144 mPa.s (25°C)
202.6 mPa.s (25 °C)
2610 mPa.s (25°C)
29.04 mPa.s (25°C)
3.291 mPa.s (25°C)
3481 mPa.s (25 °C)
392 mPa.s (25°C)
40049 mPa.s (25°C)
48.59 mPa.s (25 °C)
488.1 mPa.s (25 °C)
55.36 mPa.s (25 °C)
7.442 mPa.s (25°C)
71.11 mPa.s (25 °C)
717.4 mPa.s (25°C)
72328 mPa.s (25°C)
796.5 mPa.s (25 °C)
8.559 mPa.s (25°C)
Unit Size:
500mL
26 product options available for selection
Product selection table with 26 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Viscosity unitSize
16010272 0.2950 mPa.s (25°C) 500mL
15427840 0.5204 mPa.s (25°C) 500mL
15417840 0.9525 mPa.s (25 °C) 500mL
11994285 2.144 mPa.s (25°C) 500mL
11999748 3.291 mPa.s (25°C) 500mL
11989748 7.442 mPa.s (25°C) 500mL
15341214 8.559 mPa.s (25°C) 500mL
12166540 14.42 mPa.s (25°C) 500mL
11919758 29.04 mPa.s (25°C) 500mL
15524156 48.59 mPa.s (25 °C) 500mL
16020272 55.36 mPa.s (25 °C) 500mL
15781718 71.11 mPa.s (25 °C) 500mL
11969758 100.7 mPa.s (25°C) 500mL
15791718 124.9 mPa.s (25 °C) 500mL
16070260 202.6 mPa.s (25 °C) 500mL
16050370 392 mPa.s (25°C) 500mL
15555806 488.1 mPa.s (25 °C) 500mL
16000280 717.4 mPa.s (25°C) 500mL
15534156 796.5 mPa.s (25 °C) 500mL
16030380 1273 mPa.s (25°C) 500mL
16080260 2610 mPa.s (25°C) 500mL
15592600 3481 mPa.s (25 °C) 500mL
16020280 10030 mPa.s (25°C) 500mL
16060380 19588 mPa.s (25°C) 500mL
16040270 40049 mPa.s (25°C) 500mL
16010380 72328 mPa.s (25°C) 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 26 options available.
26 options
This item is not returnable. View return policy
Product Code. 16010272 Supplier Paragon Scientific Supplier No. N.4
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Certified in strict accordance with ASTM D2162, the primary method for viscosity standards manufacture. Dual certified to the International Standards BS EN ISO / IEC 17025 and ISO 17034 under our UKAS accrediatation.

We test at Primary level using master viscometers in accordance with ASTM D2162 to ensure the lowest uncertainty of measurement thus providing standards that are certified with primary level accuracy. ASTM D2162 is the only referenced method for the certification of viscosity standards when adopting ASTM test method protocol.

  • Certified according to ISO 17025 / ISO 17034 under our UKAS accreditation
  • Certified in strict accordance with ASTM D2162
  • Density g/mL in accordance with ASTM D1480
  • For use with glass capillary viscometers, automatic viscometers and other viscosity measuring equipment
  • Kinematic (cSt) and dynamic viscosity (mPa·s) given at all temperatures
  • Standard Pack Size 500mL
  • Fully traceable to National Standards
  • Ensures full compliance to ASTM and IP test method protocol
  • Tamper evident security packaging
  • 2 Year shelf life
  • Manufactured in the United Kingdom

Specifications

Viscosity 0.2950 mPa.s (25°C)
Quantity 500 mL
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • general perpose viscosity standard; UKAS ISO/IEC17025 and ISO guide 34 certified
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
Supplemental information
  • MIXTURE LIST-Contain: n-hexane

Nominal values quoted

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.