missing translation for 'onlineSavingsMsg'
Learn More
Learn More
- Home
- Products
- Buffers and Standards
- Viscosity Standards
- General Purpose Viscosity Standards, Paragon Scientific™
General Purpose Viscosity Standards, Paragon Scientific™
Product Code. 16070260
Change view
Click to view available options
Viscosity:
0.2950 mPa.s (25°C)
0.5204 mPa.s (25°C)
0.9525 mPa.s (25 °C)
100.7 mPa.s (25°C)
10030 mPa.s (25°C)
124.9 mPa.s (25 °C)
1273 mPa.s (25°C)
14.42 mPa.s (25°C)
19588 mPa.s (25°C)
2.144 mPa.s (25°C)
202.6 mPa.s (25 °C)
2610 mPa.s (25°C)
29.04 mPa.s (25°C)
3.291 mPa.s (25°C)
3481 mPa.s (25 °C)
392 mPa.s (25°C)
40049 mPa.s (25°C)
48.59 mPa.s (25 °C)
488.1 mPa.s (25 °C)
55.36 mPa.s (25 °C)
7.442 mPa.s (25°C)
71.11 mPa.s (25 °C)
717.4 mPa.s (25°C)
72328 mPa.s (25°C)
796.5 mPa.s (25 °C)
8.559 mPa.s (25°C)
Unit Size:
500mL
26 product options available for selection
| Product Code. | Viscosity | Unit Size |
|---|---|---|
| 16070260 | 202.6 mPa.s (25 °C) | 500mL |
| 16010272 | 0.2950 mPa.s (25°C) | 500mL |
| 15427840 | 0.5204 mPa.s (25°C) | 500mL |
| 15417840 | 0.9525 mPa.s (25 °C) | 500mL |
| 11994285 | 2.144 mPa.s (25°C) | 500mL |
| 11999748 | 3.291 mPa.s (25°C) | 500mL |
| 11989748 | 7.442 mPa.s (25°C) | 500mL |
| 15341214 | 8.559 mPa.s (25°C) | 500mL |
| 12166540 | 14.42 mPa.s (25°C) | 500mL |
| 11919758 | 29.04 mPa.s (25°C) | 500mL |
| 15524156 | 48.59 mPa.s (25 °C) | 500mL |
| 16020272 | 55.36 mPa.s (25 °C) | 500mL |
| 15781718 | 71.11 mPa.s (25 °C) | 500mL |
| 11969758 | 100.7 mPa.s (25°C) | 500mL |
| 15791718 | 124.9 mPa.s (25 °C) | 500mL |
| 16050370 | 392 mPa.s (25°C) | 500mL |
| 15555806 | 488.1 mPa.s (25 °C) | 500mL |
| 16000280 | 717.4 mPa.s (25°C) | 500mL |
| 15534156 | 796.5 mPa.s (25 °C) | 500mL |
| 16030380 | 1273 mPa.s (25°C) | 500mL |
| 16080260 | 2610 mPa.s (25°C) | 500mL |
| 15592600 | 3481 mPa.s (25 °C) | 500mL |
| 16020280 | 10030 mPa.s (25°C) | 500mL |
| 16060380 | 19588 mPa.s (25°C) | 500mL |
| 16040270 | 40049 mPa.s (25°C) | 500mL |
| 16010380 | 72328 mPa.s (25°C) | 500mL |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 26 options available.
This item is not returnable.
View return policy
Product Code.
16070260
Supplier
Paragon Scientific
Supplier No.
N100
You must Sign In and have a business account to purchase this item.
Restricted Item
In Stock
Log in to see stock.
Log in to see stock.
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
This item is not returnable.
View return policy
Description
- Certified according to ISO 17025 / ISO 17034 under our UKAS accreditation
- Certified in strict accordance with ASTM D2162
- Density g/mL in accordance with ASTM D1480
- For use with glass capillary viscometers, automatic viscometers and other viscosity measuring equipment
- Kinematic (cSt) & dynamic viscosity (mPa·s) given at all temperatures
- Standard Pack Size 500mL
- Fully traceable to National Standards
- Ensures full compliance to ASTM & IP test method protocol
- Tamper evident security packaging
- 2 Year shelf life
- Manufactured in the United Kingdom
Specifica
Specifica
| Viscosity | 202.6 mPa.s (25 °C) |
| Certifications/Compliance | ISO 17025, UKAS ISO 17034 |
| Quantity | 500 mL |
Nominal values quoted
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?