missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Geneticin™ Selective Antibiotic (G418 Sulfate), Powder

Product Code. 11538676
Click to view available options
Quantity:
10 g
Unit Size:
10g
This item is not returnable. View return policy

Product Code. 11538676

Brand: Gibco™ 11811064

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Geneticin™ Selective Antibiotic (G418 Sulfate), Powder

Geneticin™ Selection Antibiotic (G418 Sulfate) is produced by the bacterium Micromonospora rhodorangea and acts by binding the ribosome, thus inhibiting protein synthesis in both prokaryotic and eukaryotic cells. Resistance to Geneticin™ Selection Antibiotic (G418 Sulfate) is conferred by the E. coli APH(3’)–I and APH(3’)–II resistance genes.

Gibco™ Geneticin™ Selection Antibiotic (G418 Sulfate) is used as a selection antibiotic in the concentration range of 100–200μg/mL for bacteria, or 200–500μg/mL for most mammalian cells. This product is supplied as a powder and should be made into a stock solution of 10–50 mg/mL in water.

Other Choices and More Information
We offer a wide range of antibiotics and antimycotics in both powder and liquid formats.

See the complete list, or find products for:
  • Contamination control
  • Eukaryotic and bacterial selection

    See recommendations for working concentrations for selection antibiotics.

    Learn more about the use of antibiotics and antimycotics in cell culture, and review guidelines for decontaminating cultures.

    Dual-Site cGMP Manufacturing and Quality Standard
    Gibco™ Geneticin™ Selection Antibiotic (G418 Sulfate) is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer powdered Gibco™ Geneticin™ Selection Antibiotic (G418 Sulfate) products made in our New York facility (Cat. Nos. 11811098, 11811031, and 11811023). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Certifications/Compliance ISO 13485, cGMP, FDA
    Content And Storage 15 to 30°C
    Description Geneticin™ Selective Antibiotic (G418 Sulfate), 10g, 15 to 30°C Storage Temperature, Antibiotic (Selective), Geneticin™ (G-418) Agent, Antibiotic (Selective) Reagent Type, 24 Months Shelf Life, Room Temperature/Ambient
    Form Powder
    Product Type Antibiotic
    For Use With (Application) Insect cell culture, mammalian cell culture, transfection, transformation, eukaryotic selection/stable cell line generation
    Quantity 10 g
    compliance-icons
    Product Identifier
    • Geneticin Selective Antibiotic (G418 Sulfate), Powder
    Signal Word
    • Danger
    Hazard Category
    • Respiratory sensitiser Category 1
    • Skin sensitiser Category 1
    Hazard Statement
    • H317-May cause an allergic skin reaction.
    • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    Precautionary Statement
    • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
    • P272-Contaminated work clothing should not be allowed out of the workplace.
    • P280-Wear protective gloves/protective clothing/eye protection/face protection.
    • P284-¢In case of inadequate ventilation! wear respiratory protection.
    • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
    • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
    • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
    • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
    • P362+P364-Take off contaminated clothing and wash it before reuse.
    • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
    Supplemental information
    • MIXTURE LIST-Contains : D-Streptamine, O-2-amino-2,7-dideoxy-D-glycero-.alp ha.-D-gluco-heptopyranosyl-(1.fwdarw. 4)-O-[3-de

    For Research Use Only. Not for use in diagnostic procedures.

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.