missing translation for 'onlineSavingsMsg'
Learn More

Genistein 4'-beta-D-glucopyranoside, TRC

Product Code. 30240875
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30240875 100 mg 100mg
30240500 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30240875 Supplier Toronto Research Chemicals Supplier No. TRCG349975100MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 152-95-4
Molecular Formula C21 H20 O10
Molecular Weight (g/mol) 432.38
Synonym 4H-1-Benzopyran-4-one, 3-[4-(β-D-glucopyranosyloxy)phenyl]-5,7-dihydroxy-, Sophoricoside (6CI,7CI,8CI), 3-[4-(β-D-Glucopyranosyloxy)phenyl]-5,7-dihydroxy-4H-1-benzopyran-4-one, 4',5,7-Trihydroxyisoflavone 4'-β-D-glucopyranoside, 5',7'-Dihydroxy-4'-glucosyloxyisoflavone, Genistein 4'-O-glucoside, Genistein, 4'-β-D-glucopyranoside
IUPAC Name 5,7-dihydroxy-3-[4-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyphenyl]chromen-4-one
SMILES OC[C@H]1O[C@@H](Oc2ccc(cc2)C3=COc4cc(O)cc(O)c4C3=O)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C21H20O10/c22-7-15-18(26)19(27)20(28)21(31-15)30-11-3-1-9(2-4-11)12-8-29-14-6-10(23)5-13(24)16(14)17(12)25/h1-6,8,15,18-24,26-28H,7H2/t15-,18-,19+,20-,21-/m1/s1
Formula Weight 432.1056
Chemical Name or Material Sophoricoside (Genistein-4'-Glucoside)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.