missing translation for 'onlineSavingsMsg'
Learn More

Genistein 4’-Beta-D-Glucuronide, TRC

Product Code. 17994555
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17994555 2.5 mg 2.5mg
17974785 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17994555 Supplier Toronto Research Chemicals Supplier No. TRCG3499902.5MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 245084-07-5
Molecular Formula C21H18O11
Molecular Weight (g/mol) 446.36
Synonym Genistein-4'-glucuronide, 4-(5,7-Dihydroxy-4-oxo-4H-1-benzopyran-3-yl)phenyl β-D-Glucopyranosiduronic Acid;
SMILES C1=CC(=CC=C1C2=COC3=CC(=CC(=C3C2=O)O)O)O[C@H]4[C@@H]([C@H]([C@@H]([C@H](O4)C(=O)O)O)O)O

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C21H18O11/c22-9-5-12(23)14-13(6-9)30-7-11(15(14)24)8-1-3-10(4-2-8)31-21-18(27)16(25)17(26)19(32-21)20(28)29/h1-7,16-19,21-23,25-27H,(H,28,29)/t16-,17-,18+,19-,21+/m0/s1
Formula Weight 446.0849
Percent Purity >95%
Chemical Name or Material Genistein 4'-beta-D-Glucuronide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.