missing translation for 'onlineSavingsMsg'
Learn More

Genistein 7-Beta-D-Glucuronide 4’-Sulfate Disodium Salt, TRC

Product Code. 17982725
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17982725 10 mg 10mg
17991045 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17982725 Supplier Toronto Research Chemicals Supplier No. TRCG34998010MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C21H16Na2O14S
Molecular Weight (g/mol) 570.39
Synonym 5-Hydroxy-4-oxo-3-[4-(sulfooxy)phenyl]-4H-1-benzopyran-7-yl β-D-Glucopyranosiduronic Acid Disodium Salt;
SMILES O=C1C2=C(O)C=C(O[C@@H]([C@@H]3O)O[C@H](C(O[Na])=O)[C@@H](O)[C@@H]3O)C=C2OC=C1C4=CC=C(OS(=O)(O[Na])=O)C=C4

Specifications

Quantity 10 mg
Chemical Name or Material Genistein 7-beta-D-Glucuronide 4'-Sulfate Disodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.