missing translation for 'onlineSavingsMsg'
Learn More

Genistein Diglucuronide, TRC

Product Code. 17939914
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17939914 2.5 mg 2.5mg
17907924 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17939914 Supplier Toronto Research Chemicals Supplier No. TRCG3500552.5MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 428438-35-1
Molecular Formula C27H26O17
Molecular Weight (g/mol) 622.49
Synonym β-D-Glucopyranosiduronic Acid 4-[7-(β-D-Glucopyranuronosyloxy)-5-hydroxy-4-oxo-4H-1-benzopyran-3-yl]phenyl
IUPAC Name (2S,3S,4S,5R,6S)-6-(4-(7-(((2S,3R,4S,5S,6S)-6-carboxy-3,4,5-trihydroxytetrahydro-2H-pyran-2-yl)oxy)-5-hydroxy-4-oxo-4H-chromen-3-yl)phenoxy)-3,4,5-trihydroxytetrahydro-2H-pyran-2-carboxylic acid
SMILES O=C1C2=C(O)C=C(O[C@@H]([C@@H]3O)O[C@H](C(O)=O)[C@@H](O)[C@@H]3O)C=C2OC=C1C4=CC=C(O[C@H]5[C@H](O)[C@@H](O)[C@H](O)[C@@H](C(O)=O)O5)C=C4

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C27H26O17/c28-12-5-10(42-27-21(35)17(31)19(33)23(44-27)25(38)39)6-13-14(12)15(29)11(7-40-13)8-1-3-9(4-2-8)41-26-20(34)16(30)18(32)22(43-26)24(36)37/h1-7,16-23,26-28,30-35H,(H,36,37)(H,38,39)/t16-,17+,18-,19+,20+,21-,22-,23+,26+,27-/m0/s1
Formula Weight 622.12
Chemical Name or Material Genistein Diglucuronide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.