missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Gentamicin (50 mg/mL)

Product Code. 11520506 Shop All Gibco Products
Change view
Click to view available options
Quantity:
100 mL
20 x 100mL
20 x 20 mL
20 mL
Unit Size:
100mL
20mL
400mL
Pack of 20
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11520506 20 mL 20mL
11530506 100 mL 100mL
11540506 20 x 20 mL Pack of 20
11550506 20 x 100mL 400mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11520506 Supplier Gibco™ Supplier No. 15750037
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Gentamicin (50 mg/mL)

Gentamicin is prepared in distilled water.

Spectrum: Gram positive and gram negative bacteria
Recommended concentration: 0.5 to 50μg/ml
Recommended storage condition: +15°C to +30°C


Use of Gentamicin
An ideal antibiotic for cell culture should have broad spectrum antibacterial and anti-mycoplasmal activity as well as freedom from cell toxicity.Gentamicin approaches these requirements. It has biological and biochemical properties, which render it superior to the use of Penicillin and Streptomycin (PS). In cell culture it is normally used at a concentration of 25-50μg/ml in growth medium but higher concentrations e.g. 200μg/ml are often used in viral studies when specimens are known to be infected.

Advantages
Broad Spectrum - wider and more effective than Penicillin/Streptomycin
As well as being active against many gram positive or gram negative organisms, Gentamicin is active against strains of Proteus and Staphylococcus which can sometimes be resistant to a PS mix and unlike PS, Gentamicin is active against Pseudomonas strains.

Anti-Mycoplasma Activity
Although not active against yeasts, moulds or protozoa, Gentamicin has been reported to be active against several but not all strains of mycoplasma.

pH Stability
Gentamicin has been shown to be stable over a pH range of 2 to 10 in Tissue Culture Media at +37°C for 15 days, unlike Streptomycin, which is rapidly destroyed by alkaline conditions and Penicillin, which is unstable at both acid and alkaline pH.

Temperature Stability
Gentamicin is stable at all temperatures and can withstand autoclaving. Thus if using autoclavable powder media, Gentamicin can be added before sterilisation. Normal autoclaving almost completely inactivates Penicillin and reduces the activity of Streptomycin by 50%.

Activity is unaffected by the presence of Serum
Compared with Pencillin, which immediately loses 30% of its activity when added to Tissue Culture Media containing serum, Gentamicin is unaffected.

Cell Tolerance
The suggested concentration is well below maximum dose tolerated by cells. Cells will tolerate in excess of 20X normal concentrations.

Stability and replication of Viruses in the presence of Gentamicin
The use of Gentamicin does not affect virus yields or plaque formation. In addition it does not affect interferon yields or assays.

Increased Virus Isolations
At present insufficient data is available to clearly demonstrate that this is the case but accumulating evidence seems to support this claim. Basically the reason is that the superior antibiotic activity of Gentamicin allows some badly infected cultures to grow and be read whereas less effective antibiotics would not control the bacterial growth and so the culture cannot be read.

Additional Use
As an additive in mycology media to prevent bacterial growth. The properties listed above make Gentamicin ideal for use in cell and tissue culture and also for transport of cell cultures and clinical specimens containing viruses or any specimens in which overgrowth by bacteria should be avoided.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO 13485, cGMP, FDA
Content And Storage 15 to 30°C
Description Gentamicin, 20mL, 50mg/mL, 15 to 30°C Storage Temperature, Antibiotic, Gentamicin Agent, Antibiotic Reagent Type, 24 Months Shelf Life, Room Temperature/Ambient, Recommended Working Concentration Ranges From 0.5 to 50μg/mL
Concentration 50 mg/mL
Form Liquid
Product Type Antibiotic
For Use With (Application) Mammalian cell culture, prevention of cell culture contamination
Quantity 20 mL

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.