missing translation for 'onlineSavingsMsg'
Learn More

TOKU-E™ Gentamicin ReadyMade™ Solution

Product Code. 30044024
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 1 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30044024 10 x 1 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30044024 Supplier TOKU-E™ Supplier No. G04610x1ML
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Gentamicin ReadyMade™ Solution is provided as a sterile-filtered solution of Gentamicin Sulfate in water.

  • CAS Number: 1405-41-0
  • Molecular Formula: C21H43N5O7
  • Molecular Weight: 477.60 g/mol
  • Storage Conditions: 2-8°C
  • Form: Liquid
  • Source: Micromonospora purpurea or M. Echinospora
  • Spectrum: Gentamicin Sulfate is a broad-spectrum antibiotic targeting Gram-positive and Gram-negative bacteria. It is effective against several strains of Mycoplasma. It also combats certain β-lactam sensitive vancomycin-resistant Enterococcus (VRE; a 'superbug')
  • Mechanism of Action: Aminoglycosides target the 30S ribosomal subunit, blocking the translocation of peptidyl-tRNA from acceptor to donor. This results in an inability to read mRNA ultimately producing a faulty or non-existent protein

Specifications

CAS 1405-41-0
Chemical Name or Material Gentamicin
Color Colorless
Content And Storage 2°C to 8°C
Description Gentamicin ReadyMade™ Solution 50 mg/mL in Water
Concentration 50 mg/mL
Form Clear, colorless solution
Product Type Antibiotic
Sterility Sterile
Sterilization Method Sterile-filtered
With Additives None
Without Additives None
Formulation Liquid in Water
For Use With (Application) Microbiology, Media Supplements, Plant Biololgy, Eukaryotic Cell Culture, Cancer Application
Product Line ReadyMade™ Solutions
Purity or Quality Grade >95%
Quantity 10 x 1 mL
Shipping Condition Ice Pack
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.