missing translation for 'onlineSavingsMsg'
Learn More

Melford Gentamicin Sulfate

Product Code. 30018300
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
100 g
Unit Size:
100g
10g
1g
25g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018300 5 g 5g
30018298 100 g 100g
30018297 10 g 10g
30018296 1 g 1g
30018299 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 30018300 Supplier Melford Supplier No. G380005.0
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Gentamicin Sulfate is an aminoglycoside antibiotic that inhibits bacterial protein synthesis by binding to the bacterial ribosome.

Gentamicin Sulfate is an aminoglycoside antibiotic that inhibits bacterial protein synthesis by binding to the bacterial ribosome. This interference disrupts the translation process, leading to the production of faulty proteins and ultimately causing bacterial cell death.

It is frequently used as a selective agent in culture media to isolate bacteria that are sensitive to this antibiotic. Its inclusion in culture medium allows for the specific growth of bacteria lacking resistance to gentamicin.

Specifications

Chemical Name or Material Gentamicin Sulfate
CAS 1405-41-0
Quantity 5 g
Molecular Formula C21H43N5O7-H2SO4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.