missing translation for 'onlineSavingsMsg'
Learn More

Gentamicin Sulfate, Antibiotic Solution, Conc: 10mg/mL, MP Biomedicals

Product Code. 11460435 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11460435 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11460435 Supplier MP Biomedicals Supplier No. 091676045
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Antibiotic solution Concentration: 10mg/mL Effective against gram-positive and gram-negative bacteria, as well as, mycoplasma Storage temperature: 15–30°C

Specifications

Type Gentamicin Sulfate
CAS 1405-41-0
Quantity 10 mL
Chemical Name or Material Gentamicin Sulfate
compliance-icons compliance-icons
Product Identifier
  • Gentamicin Sulfate Reagent Solution
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P321-Specific treatment (see on this label).
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: gentamycin sulphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.