missing translation for 'onlineSavingsMsg'
Learn More

Gentamicin Sulfate, MP Biomedicals™

Product Code. 11480772 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11480772 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11480772 Supplier MP Biomedicals Supplier No. 02194530.6
  • ONLINE EXCLUSIVE PRICE
    47.65 €
    45.15 € / 50mg
    Save 2.50 €
    5% Off
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Inhibits the growth of a wide variety of Gram-positive and -negative microorganisms. Gentamicin Sulfate, MP Biomedicals is a broad-spectrum aminoglycoside antibiotic complex. It inhibits bacterial protein biosynthesis by binding to the 30S subunit of the

Gentamicin Sulfate is produced by fermentation of Micromonospora purpurea or M. echinospora. It is a mixture of 3 major components designated as C1, C1a, and C2. Each component consists of five basic nitrogens and requires five equivalents of sulfuric acid per mole of gentamicin base. Gentamicin is used as the sulfate salt.

Gentamicin sulfate is a broad spectrum antibiotic effective against Gram-positive and Gram-negative bacteria. It is effective against strains that are resistant to tetracycline, chloramphenicol, kanamycin and colistin, particularly strains of Pseudomonas, Proteus, Staphylococcus, and Streptococcus.

Mode of action:

  • Gentamicin causes codon misreading by binding to the 30S ribosomal subunit
  • This binding blocks the translocation of peptidyl-tRNA from the acceptor site to the donor site
  • The bactericidal effect on Pseudomonas aeruginosa is exerted by the binding of gentamicin to the outer membrane, where it displaces natural cations, destabilizes the membrane, and forms holes in the cell surface.

Specifications

Physical Form Powder
CAS 1405-41-0
pH 3.5 to 5.5 (4% solution aq. soln.) (literature)
Melting Point 218°C to 237°C (literature)
Potency >590μg/mg
Quantity 50 mg
Optical Rotation 116°
Synonym Garamycin, Genoptic, GM Sulfate;Gentiomycin C
Solubility Information Soluble in water (50mg/mL, clear to very slightly hazy, colorless to faint). Practically insoluble in alcohol and other organic solvents.
Grade Cell Culture
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.