missing translation for 'onlineSavingsMsg'
Learn More

Gentamicin Sulfate, MP Biomedicals™

Product Code. 17680887 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17680887 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17680887 Supplier MP Biomedicals Supplier No. 02190057.2
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Produced by fermentation of Micromonospora purpurea or M. echinospora. MP Biomedicals' Gentamicin Sulfate is an aminoglycoside antibiotic complex used as a selection agent in molecular biology applications. It is also used to treat many types of bacterial

The product is a mixture of 3 major components designated as C1, C1a, and C2. Gentamicin is used as the sulfate salt. Each component consists of five basic nitrogens and requires five equivalents of sulfuric acid per mole of gentamicin base.

  • Used as a selection agent in molecular biology applications
  • Treats many types of bacterial infections, particularly those caused by Gram-negative organisms

Specifications

Physical Form Powder
CAS 1405-41-0
pH 3.5 to 5.5 (1 in 25, aq. soln.)
Melting Point 218°C to 237°C (as powder)(literature)
Ignition Residue ≤1%
Loss on Drying ≤18%
Potency ≤590μg/mg
Quantity 250 mg
Optical Rotation 107–121° (10mg/mL, water)
Synonym Garamycin, Gentamicin sulfate salt, Gentiomycin C
Solubility Information H2O: soluble 50mg/mL
Grade USP
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Gentamycin Sulfate, USP
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: gentamycin sulphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.