Learn More
Invitrogen™ Gephyrin Monoclonal Antibody (24GB5585)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA557412
 View more versions of this product
Product Code. 30276281
  • 564.00 € / 100µL
Description
Gephyrin Monoclonal Antibody for Western Blot, Flow
Gephyrin is a neuronal assembly protein that anchors inhibitory neurotransmitter receptors to the postsynaptic cytoskeleton via high affinity binding to a receptor subunit domain and tubulin dimers. In nonneuronal tissues, the encoded protein is also required for molybdenum cofactor biosynthesis. Mutations in this gene may be associated with the neurological condition hyperplexia and also lead to molybdenum cofactor deficiency. Numerous alternatively spliced transcript variants encoding different isoforms have been described; however, the full-length nature of all transcript variants is not currently known.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Gephyrin
Monoclonal
Unconjugated
Gphn
5730552E08Rik; AI662856; BC027112; C230040D23; Domain E; Domain G; EGK_18299; Geph; gephyrin; gephyrin isoform; Gph; GPHN; GPHRYN; HKPX1; KIAA1385; MOCODC; Molybdopterin adenylyltransferase; Molybdopterin molybdenumtransferase; MPT adenylyltransferase; MPT Mo-transferase; Putative glycine receptor-tubulin linker protein
Mouse
Affinity chromatography
RUO
10243, 268566
-20°C
Liquid
Flow Cytometry, Western Blot
24GB5585
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q8BUV3, Q9NQX3
Gphn
Recombinant protein of human GPHN (aa 1-736).
100 μL
Primary
Human, Mouse
Antibody
IgG2a