missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Germanium Standard For ICP, 1000μg/mL in H2O, Assurance Grade, SPEX CertiPrep™
Shop All SPEX Certiprep Products
Click to view available options
Quantity:
125 mL
500 mL
Packaging:
Triple-leached LDPE bottle
Unit Size:
125mL
500mL
Description
Includes
NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| Ge | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Specifications
| Packaging | Triple-leached LDPE bottle |
| Certificate of Analysis | Assurance Certificate of Analysis reports up to 39 elements at ppb levels. |
| Concentration or Composition Notes | Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. |
| Technique | AA, ICP |
| For Use With (Equipment) | AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation |
| Type | Certified Reference Material |
| Solvent or Matrix | Water/Trace F- |
| Quantity | 500 mL |
| Grade | Assurance |
| Concentration or Composition (by Analyte or Components) | Matrix Concentration Cat. No. H2O/tr. HF 1000μg/mL PLGE9-2Y, -2X 10,000 PLGE9-3Y, -3X |
| Show More |
Product intended for laboratory use only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto