missing translation for 'onlineSavingsMsg'
Learn More

Germanium Standard: Ge @ 10000 μg/mL in 5% HNO3, tr. HF, VHG™

Product Code. 30003177
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Unit Size:
100mL
250mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30003177 250 mL 250mL
30003098 500 mL 500mL
30002984 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 30003177 Supplier VHG Labs Supplier No. VHGTGENF250
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Germanium Standard: Ge @ 10000 μg/mL in 5% HNO3, tr. HF
TRUSTED_SUSTAINABILITY

Specifikationer

Chemical Name or Material Germanium
CAS 7440-56-4
Methods ICP/ICP-MS
Analyte or Component Names Germanium
Shipping Condition Room Temperature
Molecular Formula Ge
Concentration 10000 μg/mL
Certifications/Compliance ISO 17034
Synonym Ge
InChI Formula InChI=1S/Ge
SMILES [Ge]
IUPAC Name germanium
Solvent or Matrix Dilute HNO3
Molecular Weight (g/mol) 72.64
Quantity 250 mL
Formula Weight 73.9212 g/mol
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.