missing translation for 'onlineSavingsMsg'
Learn More

Gestodene, MedChemExpress

Product Code. 16484237
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16484237 50 mg 50mg
16428106 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16484237 Supplier Medchem Express Supplier No. HYB0110
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Gestodene(SHB 331;WL 70) is a progestogen hormonal contraceptive.

Gestodene(SHB 331

Specifications

Chemical Name or Material Gestodene
Physical Form Solid
CAS 60282-87-3
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C21H26O2
Synonym SHB 331 WL 70
Solubility Information DMSO : ≥ 100 mg/mL (322.13 mM)
SMILES CC[C@@]1([C@]2(O)C#C)[C@](C=C2)([H])[C@@](CCC3=CC4=O)([H])[C@]([C@@]3([H])CC4)([H])CC1
Molecular Weight (g/mol) 310.43
Formula Weight 310.43
Percent Purity 99.69%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.