missing translation for 'onlineSavingsMsg'
Learn More

GFP Antibody, FITC, R&D Systems™

Product Code. 18097074 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
Unit Size:
25µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18097074 0.025 mg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18097074 Supplier Novus Biologicals Supplier No. NB10011830.025MG
  • ONLINE EXCLUSIVE PRICE
    274.19 €
    232.05 € / 25µg
    Save 42.14 €
    15% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

GFP Polyclonal specifically detects GFP in Non-species specific samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GFP
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Conjugate FITC
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1:2000-1:10000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:500-1:2500, Immunohistochemistry-Frozen
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Sodium Azide
Gene Accession No. P42212
Gene Alias Enhanced Green Fluorescent Protein, Green Fluorescent Protein, green fluorescent protein (gfp)
Gene Symbols GFP
Host Species Rabbit
Immunogen Highly purified recombinant full length protein made in E. coli. The antibody is directed against the entire GFP molecule.
Molecular Weight of Antigen 27 kDa
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity GFP from Aequorea victoria
Target Species Non-species specific
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.