missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GFRAL Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5107133

Product Code. 17116943

  • 592.00 € / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Sequence homology: Cow: 85%; Human: 100%; Pig: 77%; Rabbit: 92%.

Brainstem-restricted receptor for GDF15 which regulates food intake, energy expenditure and body weight in response to metabolic and toxin-induced stresses (PubMed:28953886, PubMed:28846097, PubMed:28846098, PubMed:28846099). Upon interaction with its ligand, GDF15, interacts with RET and induces cellular signaling through activation of MAPK- and AKT- signaling pathways. [UniProt].
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GFRAL
Polyclonal
Unconjugated
GFRAL
AY457637; bA360D14.1; C6orf144; GDNF family receptor alpha like; GDNF family receptor alpha-like; GDNF receptor alpha-like; GFRAL; Gral; IVFI9356; RGD1565220; UNQ9356; UNQ9356/PRO34128
Rabbit
Affinity Chromatography
RUO
389400
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide; pH 7.4
Q6UXV0
GFRAL
Synthetic peptide directed towards the N-terminal region of Human GFRAL.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.