missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GGH Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13264-1-AP-150UL
This item is not returnable.
View return policy
Description
Gamma-Glutamyl hydrolase (GGH) is a homodimeric protein that catalyzes the cleavage of the gamma-glutamyl chain of folylpoly-gamma-glutamyl substrates and is a central enzyme in folyl and antifolyl poly-gamma-glutamate metabolism. The activities of the baculovirus-expressed enzyme and the enzyme from HL-60 cells were similar on methotrexate polyglutamates. GGH is expressed in a variety of human tuomrs and correlates with poor prognosis.Specifications
| GGH | |
| Polyclonal | |
| Unconjugated | |
| GGH | |
| Conjugase, Gamma Glu X carboxypeptidase, Gamma glutamyl hydrolase, GGH, GH | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14590, 25455, 8836 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q62867, Q92820, Q9Z0L8 | |
| GGH | |
| GGH Fusion Protein Ag3939 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction